COM448687574 Combi-Blocks, Inc.

N-(2-amino-5,7-dibromo-1,3-benzoxazol-4-yl)-2,2-dimethylpropanamide

  • MDL number MFCD27936898
  • Cas Number : 1820704-83-3
  • Molecular Weight : 391.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3099
Availability details for
Quantity
  • Availability
  • Add to Cart