COM448687575 Combi-Blocks, Inc.

4-Chloro-5-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD28101660
  • Cas Number : 1820717-21-2
  • Molecular Weight : 182.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3100
Availability details for
Quantity
  • Availability
  • Add to Cart