COM448687579 Combi-Blocks, Inc.

2-amino-5-fluoro-1,3-benzoxazole-7-carboxylic acid

  • MDL number MFCD27936902
  • Cas Number : 1820684-43-2
  • Molecular Weight : 196.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3104

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3104
Availability details for
Quantity
  • Availability
  • Add to Cart