COM448687581 Combi-Blocks, Inc.

2-amino-6-chloro-1,3-benzoxazol-5-ol

  • MDL number MFCD27936904
  • Cas Number : 1820717-24-5
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3106
Availability details for
Quantity
  • Availability
  • Add to Cart