COM448687584 Combi-Blocks, Inc.

6-bromo-7-fluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD27936906
  • Cas Number : 1820674-33-6
  • Molecular Weight : 231.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3109

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3109
Availability details for
Quantity
  • Availability
  • Add to Cart