COM448687587 Combi-Blocks, Inc.

6-bromo-4-fluoro-1,3-benzoxazol-2-amine

  • MDL number MFCD27936909
  • Cas Number : 1820711-90-7
  • Molecular Weight : 231.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3112
Availability details for
Quantity
  • Availability
  • Add to Cart