COM448687590 Combi-Blocks, Inc.

methyl 2-amino-6-fluoro-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD27936912
  • Cas Number : 1820665-90-4
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3115
Availability details for
Quantity
  • Availability
  • Add to Cart