COM448687591 Combi-Blocks, Inc.

2-amino-[1,3]oxazolo[4,5-g]quinazolin-8-ol

  • MDL number MFCD28166425
  • Cas Number : 1820717-54-1
  • Molecular Weight : 202.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3116
Availability details for
Quantity
  • Availability
  • Add to Cart