COM448687593 Combi-Blocks, Inc.

2-amino-5,6,7-trifluoro-1,3-benzoxazole-4-carboxylic acid

  • MDL number MFCD27936914
  • Cas Number : 1820612-84-7
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3118

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3118
Availability details for
Quantity
  • Availability
  • Add to Cart