COM448687596 Combi-Blocks, Inc.

6-fluoro-1,3-benzoxazole-2,4-diamine

  • MDL number MFCD27936917
  • Cas Number : 1820619-85-9
  • Molecular Weight : 167.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3121
Availability details for
Quantity
  • Availability
  • Add to Cart