COM448687607 Combi-Blocks, Inc.

2-amino-7-methoxy-1,3-benzoxazole-5-carbonitrile

  • MDL number MFCD28166433
  • Cas Number : 1820735-51-0
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3132
Availability details for
Quantity
  • Availability
  • Add to Cart