COM448687609 Combi-Blocks, Inc.

5-nitro-[1,3]oxazolo[4,5-h]quinolin-2-amine

  • MDL number MFCD28166435
  • Cas Number : 1820620-36-7
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3134
Availability details for
Quantity
  • Availability
  • Add to Cart