COM448687613 Combi-Blocks, Inc.

2-amino-1,3-benzoxazole-7-carbaldehyde

  • MDL number MFCD28166438
  • Cas Number : 1804411-34-4
  • Molecular Weight : 162.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3138
Availability details for
Quantity
  • Availability
  • Add to Cart