COM448687616 Combi-Blocks, Inc.

ethyl 2-amino-7-bromo-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD27936922
  • Cas Number : 1820687-34-0
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3141
Availability details for
Quantity
  • Availability
  • Add to Cart