COM448687621 Combi-Blocks, Inc.

(2-amino-1,3-benzoxazol-5-yl)acetic acid

  • MDL number MFCD28166445
  • Cas Number : 1453089-83-2
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3146

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3146
Availability details for
Quantity
  • Availability
  • Add to Cart