COM448687622 Combi-Blocks, Inc.

methyl 2-amino-6-hydroxy-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD28166446
  • Cas Number : 1820666-51-0
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3147
Availability details for
Quantity
  • Availability
  • Add to Cart