COM448687624 Combi-Blocks, Inc.

2-amino-7-propoxy-1,3-benzoxazole-5-carbaldehyde

  • MDL number MFCD28166448
  • Cas Number : 1820687-19-1
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3149
Availability details for
Quantity
  • Availability
  • Add to Cart