COM448687625 Combi-Blocks, Inc.

4-(trifluoromethyl)-1,3-benzoxazol-2-amine

  • MDL number MFCD28166449
  • Cas Number : 1806424-43-0
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3150
Availability details for
Quantity
  • Availability
  • Add to Cart