COM448687631 Combi-Blocks, Inc.

8-methyl-5-nitro-[1,3]oxazolo[4,5-h]quinolin-2-amine

  • MDL number MFCD28166453
  • Cas Number : 1820650-83-6
  • Molecular Weight : 244.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3156
Availability details for
Quantity
  • Availability
  • Add to Cart