COM448687632 Combi-Blocks, Inc.

methyl 2-amino-6-chloro-1,3-benzoxazole-5-carboxylate

  • MDL number MFCD28166454
  • Cas Number : 1820704-00-4
  • Molecular Weight : 226.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3157
Availability details for
Quantity
  • Availability
  • Add to Cart