COM448687633 Combi-Blocks, Inc.

6,7-difluoro-1,3-benzoxazol-2-amine hydrochloride

  • MDL number MFCD27936924
  • Cas Number : 1820608-43-2
  • Molecular Weight : 206.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3158
Availability details for
Quantity
  • Availability
  • Add to Cart