COM448687634 Combi-Blocks, Inc.

1-(2-amino-1,3-benzoxazol-7-yl)ethanone hydrochloride

  • MDL number MFCD27936925
  • Cas Number : 1820615-08-4
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3159
Availability details for
Quantity
  • Availability
  • Add to Cart