COM448687635 Combi-Blocks, Inc.

2-amino-N,N-dimethyl-1,3-benzoxazole-6-sulfonamide

  • MDL number MFCD27936926
  • Cas Number : 1820605-24-0
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3160
Availability details for
Quantity
  • Availability
  • Add to Cart