COM448687636 Combi-Blocks, Inc.

1-[2-amino-4-(benzyloxy)-1,3-benzoxazol-7-yl]ethanone hydrochloride

  • MDL number MFCD28166455
  • Cas Number : 1820684-13-6
  • Molecular Weight : 318.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3161

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3161
Availability details for
Quantity
  • Availability
  • Add to Cart