COM448687637 Combi-Blocks, Inc.

7-chloro-5-nitro-1,3-benzoxazol-2-amine

  • MDL number MFCD28166456
  • Cas Number : 1820717-50-7
  • Molecular Weight : 213.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3162
Availability details for
Quantity
  • Availability
  • Add to Cart