COM448687645 Combi-Blocks, Inc.

naphtho[2,1-d][1,3]oxazol-2-amine hydrochloride

  • MDL number MFCD28166464
  • Cas Number : 1820734-64-2
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3170
Availability details for
Quantity
  • Availability
  • Add to Cart