COM448687649 Combi-Blocks, Inc.

methyl 2-(2-amino-1,3-benzoxazol-5-yl)acetate

  • MDL number MFCD28166468
  • Cas Number : 1781911-47-4
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3174
Availability details for
Quantity
  • Availability
  • Add to Cart