COM448687650 Combi-Blocks, Inc.

5-bromo-1,3-benzoxazol-2-amine hydrochloride

  • MDL number MFCD28166469
  • Cas Number : 1820674-67-6
  • Molecular Weight : 249.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3175
Availability details for
Quantity
  • Availability
  • Add to Cart