COM448687655 Combi-Blocks, Inc.

2-amino-N-methyl-1,3-benzoxazole-5-sulfonamide

  • MDL number MFCD28166474
  • Cas Number : 1820651-27-1
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3180
Availability details for
Quantity
  • Availability
  • Add to Cart