COM448687656 Combi-Blocks, Inc.

5-(trifluoromethyl)-1,3-benzoxazol-2-amine hydrochloride

  • MDL number MFCD28166475
  • Cas Number : 1820707-59-2
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-3181
Availability details for
Quantity
  • Availability
  • Add to Cart