COM448687660 Combi-Blocks, Inc.

(4-Cyano-2-nitrophenyl)malonic acid

  • MDL number MFCD08436992
  • Cas Number : 927801-05-6
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5005
Availability details for
Quantity
  • Availability
  • Add to Cart