COM448687662 Combi-Blocks, Inc.

2-(4-Methanesulfonyl-2-nitrophenyl)propanedioic acid

  • MDL number MFCD06203246
  • Cas Number : 773869-70-8
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5007
Availability details for
Quantity
  • Availability
  • Add to Cart