COM448687663 Combi-Blocks, Inc.

2-[2-nitro-4-(trifluoromethyl)phenyl]propanedioic acid

  • MDL number MFCD27578959
  • Cas Number : 1820640-28-5
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5008
Availability details for
Quantity
  • Availability
  • Add to Cart