COM448687665 Combi-Blocks, Inc.

4-Methoxy-3-oxobutanoic acid

  • MDL number MFCD19228814
  • Cas Number : 68665-45-2
  • Molecular Weight : 132.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5010
Availability details for
Quantity
  • Availability
  • Add to Cart