COM448687666 Combi-Blocks, Inc.

norleucine hydrochloride

  • MDL number MFCD27578961
  • Cas Number : 70288-84-5
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5012
Availability details for
Quantity
  • Availability
  • Add to Cart