COM448687667 Combi-Blocks, Inc.

3-[(carboxymethyl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD27578962
  • Cas Number : 1820607-57-5
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5028
Availability details for
Quantity
  • Availability
  • Add to Cart