COM448687669 Combi-Blocks, Inc.

1-[(tert-butoxy)carbonyl]-2-(dihydroxyboranyl)indole-7-carboxylic acid

  • MDL number MFCD27578965
  • Molecular Weight : 305.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5035
Availability details for
Quantity
  • Availability
  • Add to Cart