COM448687674 Combi-Blocks, Inc.

3-Methoxycarbonylmethoxy-4-nitro-benzoic acid methyl ester

  • MDL number MFCD13704918
  • Cas Number : 138035-71-9
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5069
Availability details for
Quantity
  • Availability
  • Add to Cart