COM448687675 Combi-Blocks, Inc.

(2S)-2-amino-3-{[(tert-butoxy)carbonyl]amino}propanoic acid hydrochloride

  • MDL number MFCD27578969
  • Cas Number : 1820572-31-3
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5072
Availability details for
Quantity
  • Availability
  • Add to Cart