COM448687677 Combi-Blocks, Inc.

(2S)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid; acetic acid

  • MDL number MFCD27578970
  • Cas Number : 42417-17-4
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5079
Availability details for
Quantity
  • Availability
  • Add to Cart