COM448687679 Combi-Blocks, Inc.

2-(Dimethoxyphosphoryl)acetic acid

  • MDL number MFCD03788193
  • Cas Number : 34159-46-1
  • Molecular Weight : 168.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5083
Availability details for
Quantity
  • Availability
  • Add to Cart