COM448687680 Combi-Blocks, Inc.

(triphenylphosphanylidene)acetic acid

  • MDL number MFCD03426562
  • Cas Number : 15677-02-8
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5088

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5088
Availability details for
Quantity
  • Availability
  • Add to Cart