COM448687685 Combi-Blocks, Inc.

(E)-3-(4-(Bromomethyl)phenyl)acrylic acid

  • MDL number MFCD07644585
  • Cas Number : 927800-43-9
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5212
Availability details for
Quantity
  • Availability
  • Add to Cart