COM448687687 Combi-Blocks, Inc.

(2S)-3-hydroxy-2-[(triphenylmethyl)amino]propanoic acid

  • MDL number MFCD27578974
  • Cas Number : 4465-45-6
  • Molecular Weight : 347.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5251
Availability details for
Quantity
  • Availability
  • Add to Cart