COM448687702 Combi-Blocks, Inc.

2-Amino-4-(4-methoxy-phenyl)-thiazole-5-carboxylic acid

  • MDL number MFCD06849881
  • Cas Number : 899350-40-4
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5362
Availability details for
Quantity
  • Availability
  • Add to Cart