COM448687704 Combi-Blocks, Inc.

7,8-Dimethyl-4-oxo-1,4-dihydro-2-quinolinecarboxylic acid

  • MDL number MFCD18343490
  • Cas Number : 52979-91-6
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5364
Availability details for
Quantity
  • Availability
  • Add to Cart