COM448687710 Combi-Blocks, Inc.

5,7-Dichlorokynurenic acid

  • MDL number MFCD00083173
  • Cas Number : 131123-76-7
  • Molecular Weight : 258.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5378
Availability details for
Quantity
  • Availability
  • Add to Cart