COM448687713 Combi-Blocks, Inc.

3-{[3-(dihydroxyboranyl)phenyl]formamido}propanoic acid

  • MDL number MFCD27578977
  • Molecular Weight : 237.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5384
Availability details for
Quantity
  • Availability
  • Add to Cart