COM448687717 Combi-Blocks, Inc.

8-bromo-4-chloro-5-fluoroquinoline-2-carboxylic acid

  • MDL number MFCD27578980
  • Cas Number : 1695285-00-7
  • Molecular Weight : 304.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5391
Availability details for
Quantity
  • Availability
  • Add to Cart