COM448687718 Combi-Blocks, Inc.

4-Hydroxy-7-(trifluoromethyl)quinoline-2-carboxylic acid

  • MDL number MFCD24387460
  • Cas Number : 36303-10-3
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YD-5392
Availability details for
Quantity
  • Availability
  • Add to Cart